missing translation for 'onlineSavingsMsg'
Learn More

Tubercidin, MedChemExpress

Product Code. 16442788
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16442788

Brand: Medchem Express HY100126

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tubercidin (7-Deazaadenosine) is an antibiotic obtained from Streptomyces tubercidicus.

Tubercidin inhibits the growth of Streptococcus faecalis (8043) with an IC50 of 0.02 μM. Tubercidin inhibits polymerases by incorporating DNA or RNA, thereby inhibiting DNA replication, RNA and protein synthesis. Tubercidin is a weak inhibitor of adenosine phosphorylase, and interferes with the phosphorylation of adenosine and AMP. Tubercidin has antiviral activity.

Specifications

Chemical Name or Material Tubercidin
Physical Form Solid
CAS 69-33-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C11H14N4O4
For Use With (Application) COVID-19-anti-virus
Synonym 7-Deazaadenosine
Solubility Information DMSO : ≥ 30 mg/mL (112.68 mM)
SMILES NC1=C2C(N([C@H]3[C@@H]([C@@H]([C@@H](CO)O3)O)O)C=C2)=NC=N1
Molecular Weight (g/mol) 266.25
Formula Weight 266.25
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.