Learn More
Benchmark Scientific™ Tube Block for Benchmark MultiTherm
Description
- Accommodates 24 x 12 mm tubes
- Protective shield protects users from burns
- Fits MultiTherm H and HC Models
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Chamber Material | Aluminum |
| Holds | 24 x 12 mm Tubes |
| Material | Aluminum |
| No. of Places | 24 |
| Type | Block |
| For Use With (Application) | Vortexing of 12 mm Diameter Tubes |