Description
- For use with UDK Series Distillation Units
Specifications
Specifications
| Length (Metric) | 260 mm |
| Type | Spacer |
| For Use With (Equipment) | Test Tube |
| Diameter (Metric) Outside | 48mm |
| Quantity | 1Each |