Learn More
Tubastatin A Hydrochloride, MedChemExpress
Product Code. 16476146
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. 16476146
Brand: Medchem Express HY13271
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Tubastatin A Hydrochloride (Tubastatin A HCl) is a potent and selective HDAC6 inhibitor with IC50 of 15 nM in a cell-free assay, and is selective (1000-fold more) against all other isozymes except HDAC8 (57-fold more). Tubastatin A Hydrochloride also inhibits HDAC10 and metallo-β-lactamase domain-containing protein 2 (MBLAC2).
Tubastatin A Hydrochloride (Tubastatin A HCl) is a potent and selective HDAC6 inhibitor with IC50 of 15 nM in a cell-free assay, and is selective (1000-fold more) against all other isozymes except HDAC8 (57-fold more). Tubastatin A Hydrochloride also inhibits HDAC10 and metallo-β-lactamase domain-containing protein 2 (MBLAC2).
Specifications
Chemical Name or Material Tubastatin A Hydrochloride
CAS 1310693-92-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H22ClN3O2
Quantity 100 mg
Synonym Tubastatin A HCl TSA HCl
Solubility Information DMSO : 10.8 mg/mL (29.04 mM; Need ultrasonic and warming) ∣H2O : 6.67 mg/mL (17.94 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C=C1)CN2C3=C(C4=C2C=CC=C4)CN(C)CC3)NO.[H]Cl
Molecular Weight (g/mol) 371.86
Formula Weight 371.86
Percent Purity 97.17%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death