missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTP Antibody (OTI8B5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-02105
This item is not returnable.
View return policy
Description
TTP Monoclonal specifically detects TTP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TTP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
| ZFP36 | |
| Full length human recombinant protein of human ZFP36 (NP_003398) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 7538 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI8B5 | |
| Unconjugated | |
| EZNFTranscription factor 17, HKL1Zinc finger protein eZNF, KID-1, KID1TCF-17, TCF17, zinc finger protein 354A | |
| Mouse | |
| 33.8 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction