missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSX Flex Series High Performance 142 L Undercounter Refrigerator

Product Code. 32237164 Shop All Thermo Scientific Products
Change view
Click to view available options
Door Style:
Glass
Solid
Voltage:
220 V
Plug Type:
CEE 7/7
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Door Style Voltage Plug Type Unit Size
32237164 Solid 220 V CEE 7/7 Each
32237163 Glass 220 V CEE 7/7 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32237164 Supplier Thermo Scientific™ Supplier No. TSX10505SV
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ TSX Series high-performance, Innovative refrigeration solutions for healthcare and research needs.

Thermo Scientific™ TSX HP Flex Undercounter Laboratory Refrigerators provide dependable laboratory cold storage in a compact footprint for space-constrained environments. Designed for the storage of reagents, media, kits, samples, vaccines, and other temperature-sensitive materials, these refrigerators provide reliable temperature performance for everyday laboratory applications. Touchscreen controls and onboard data access simplify everyday laboratory operation.

Compact performance for space-constrained laboratories

  • Designed for the storage of reagents, media, kits, samples, vaccines, and other temperature-sensitive laboratory materials
  • Compact undercounter footprint maximizes valuable laboratory workspace while delivering dependable cold storage performance
  • Ideal for research, clinical, diagnostic, pharmaceutical, biotechnology, industrial, and academic laboratory environments
  • Designed to support convenient access to stored laboratory materials in point-of-use applications

Designed for flexible laboratory layouts

  • Fits beneath standard laboratory work surfaces to maximize available floor space
  • Stackable refrigerator and freezer configurations provide additional storage capacity within a single footprint
  • Consistent user interface and control logic across the TSX portfolio simplifies training and day-to-day operation
  • Uses the same proven TSX control architecture while utilizing a streamlined control interface

Reliable performance for everyday laboratory workflows

  • Engineered to deliver dependable temperature control for routine laboratory applications
  • Designed to provide reliable point-of-use storage for commonly used laboratory materials
  • Built on the trusted TSX platform for long-term performance, usability, and reliability
  • Designed to support dependable cold storage performance and efficient everyday operation

Designed for confidence at the point of use

  • Modern touchscreen interface simplifies daily operation, configuration, and status review
  • Onboard data logging with USB download provides convenient access to temperature records for monitoring and documentation
  • Intuitive controls support faster training, easier setup, and greater confidence during everyday operation

Compliance-ready cold storage

  • Designed to support qualification, validation, and audit-ready laboratory environments
  • cULus or CE listed, as appropriate by voltage and region
  • REACH, RoHS, Proposition 65, and WEEE compliant
  • North American 115V / 60Hz models are ENERGY STAR™ 2.0 certified, where applicable

Sustainable by design

  • Variable-speed compressor technology adapts cooling performance based on laboratory demand while maintaining stable temperatures
  • Designed to support applicable EU F-Gas regulations and the U.S. EPA SNAP program
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 5 cu.ft.
Capacity (Metric) 142 L
Temperature Range 2°C to 8°C
Door Style Solid
No. of Doors 1
Exterior Finish Powder-coated Steel
Certifications/Compliance CE, cULus, NSF 456, Prop 65, REACH
Style Undercounter
Control Type Touch Screen
Warranty 5 years parts and labor, plus an additional 5 years compressor parts coverage
Defrost Automatic
Lighting LED
Access Security Key Lock
Data Outputs USB
Compressor V-drive
Dimensions (D x W x H) Interior 465 x 505 x 595 mm
Dimensions (L x W x H) Exterior 670.5 x 607 x 841 mm
Plug Type CEE 7/7
Voltage 220 V
Frequency 50/60Hz
Weight (English) 132 lb.
Weight (Metric) 60 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.