Learn More
TSPYL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA14261
Detalles adicionales : Weight : 0.01000kg
Descripción
PA1-4261 detects the DENTT protein in human samples. PA1-4261 has been successfully used Western blot procedures. PA1-4261 immunizing peptide corresponds to amino acid residues 489-503 of the human DENTT protein.
This gene encodes the beta-subunit of glucosidase II, an N-linked glycan-processing enzyme in the endoplasmic reticulum (ER). This protein is an acidic phospho-protein known to be a substrate for protein kinase C. Mutations in this gene have been associated with the autosomal dominant polycystic liver disease (PCLD). Alternatively spliced transcript variants encoding distinct isoforms have been observed.Especificaciones
TSPYL2 | |
Polyclonal | |
Unconjugated | |
TSPYL2 | |
CASK interacting nucleosome assembly protein; CASK-interacting nucleosome assembly protein; CDA1; Cell division autoantigen 1; cell division autoantigen 1 nucleolar protein; CINAP; CTCL; CTCL tumor antigen se20-4; CTCL-associated antigen se20-4; Cutaneous T-cell lymphoma-associated antigen se20-4; cutaneous T-cell lymphoma-associated tumor antigen se20-4; DENTT; differentially expressed nucleolar TGF-beta1 target; differentially-expressed nucleolar TGF-beta1 target protein; DXBwg1396e; DXHXS1008E; E130307F10Rik; HRIHFB2216; NP79; nuclear protein of 79 kDa; nucleolar TGF-beta1 target protein; RGD1563283; RP1-290F12.2; SE204; testis-specific protein Y encoded-like 2; testis-specific Y-encoded-like protein 2; TSPX; TSPY like 2; Tspyl2; TSPY-like 2; TSPY-like protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64061 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9H2G4 | |
TSPYL2 | |
Synthetic peptide: N(489) N E T T D N N E S A D D H E(503) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |