missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSH beta Antibody (TSH-116) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-363
This item is not returnable.
View return policy
Description
TSH beta Monoclonal specifically detects TSH beta in Human samples. It is validated for ELISA, Radioimmunoassay.
Specifications
| Thyroid Stimulating Hormone | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1:2000, Radioimmunoassay 1:100-1:2000 | |
| TSHB | |
| Human thyrotropin. | |
| 0.1 mg | |
| Cancer | |
| 7252 | |
| Human | |
| IgG2a |
| ELISA, Radioimmune Assays (RIA) | |
| TSH-116 | |
| Unconjugated | |
| thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody (clone TSH-116) reacts with human thyroid stimulating hormone (hTSH, thyrotropin), a glycoprotein hormone produced by the anterior pituitary gland cells in response to signals from the hypothalamus gland in the brain. The TSH-116 reacts with association constant 1.1 x 1011 l/mol. Following cross-reactivity expressed as binding of labelled hormone (% of total) was determined by solid phase RIA with excess of the antibody TSH-116: hTSH (78.9), hCG (20.3), hLH (23.2), hFSH (29.9). | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction