missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSFM Polyclonal Antibody
GREENER_CHOICE

Product Code. 15574252
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15574252 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15574252 Supplier Invitrogen™ Supplier No. PA540581

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: KQAEIWLHKE AQKEGWSKAA KLQGRKTKEG LIGLLQEGNT TVLVEVNCET Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%; Zebrafish: 77%.

Synthesis of the 13 mitochondrial-encoded proteins occurs on a dedicated mitochondrial translation apparatus similar to that found in prokaryotes and requires, in addition to the tRNAs and rRNAs encoded in mtDNA, the concerted action of several translation factors and a large number of mitochondrial ribosomal proteins, all of which are encoded by nuclear genes. The TSFM gene encodes a mitochondrial translation elongation factor (Smeitink et al., 2006 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSFM
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TSFM
Gene Accession No. P43897
Gene Alias 2310050B20Rik; 2A3-2; 9430024O13Rik; EFTS; EF-Ts; EF-Ts {ECO:0000255; EFTSMT; EF-TsMt; EF-TsMt {ECO:0000255; elongation factor Ts; elongation factor Ts, mitochondrial; elongation factor Ts, mitochondrial {ECO:0000255; HAMAP-Rule:MF_03135}; mitochondrial elongation factor Ts; translation elongation factor; Ts translation elongation factor, mitochondrial; TSFM; zgc:158429
Gene Symbols TSFM
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TSFM (aa 76-125).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10102
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.