missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSCOT Polyclonal Antibody
GREENER_CHOICE

Product Code. 13445907
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13445907 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13445907 Supplier Invitrogen™ Supplier No. PA531389

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A549, H1299, HCT116, MCF-7. Predicted reactivity: Dog (84%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The TSCOT gene, known as the Thymic Stromal Co-transporter, plays a crucial role in the gene expression regulation within thymic epithelial cells, essential for T cell development. It encodes a protein that bears weak homology to bacterial 12 transmembrane co-transporters. Research indicates that TSCOT is predominantly expressed in thymic tissues, although low levels of expression are detectable in other epithelial tissues such as skin and lungs. The gene has been co-analyzed with Foxn1 and Hoxa3, highlighting its significant role in organogenesis. Studies utilizing competitive reverse transcription-polymerase chain reaction (RT-PCR) have mapped TSCOT to specific genomic regions, verifying its expression in a tissue and cell-type specific manner. The introduction of a neoantigen into TSCOT-expressing cells demonstrates its capability to mediate antigen-specific T cell tolerance, offering potential applications in immunological therapies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSCOT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SLC46A2
Gene Accession No. Q9BY10
Gene Alias 5430429N04Rik; Ly110; putative thymic stromal co-transporter TSCOT; SLC46A2; Solute carrier family 46 member 2; solute carrier family 46, member 2; thymic stromal co-transporter; thymic stromal cotransporter homolog; Thymic stromal cotransporter protein; TSCOT; TSO-1C12
Gene Symbols SLC46A2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TSCOT. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57864
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.