missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tryptone Soya Agar with 1% Tween
Click to view available options
Fill Weight:
500 to 504 g
Unit Size:
Each
Description
Tryptone Soya Agar with 1% Tween is for recovery and enumeration of microorganisms from surfaces, materials, or products, especially after cleaning or disinfection procedures.
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2°C to 25°C |
| Format | 500 mL Sirop Bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 500 to 504 g |
| Packaging Type | 10 Sirop Bottles with screw cap closures in a box |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction