Learn More
STEMCELL Technologies Trypsin-EDTA (0.25%)
Product Code. 17129901
Encompass_Preferred
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 17129901
Brand: STEMCELL Technologies 07901
Please to purchase this item. Need a web account? Register with us today!
Enzymatic cell dissociation reagent
  • Trypsin-EDTA (0.25%) is recommended for the dissociation of adherent cells, cell aggregates, and tissues into single-cell suspensions
Specifications
Quantity 500 mL