missing translation for 'onlineSavingsMsg'
Learn More

TRPC5 Antibody (N67/15), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-12919-0.025mg

 View more versions of this product

Product Code. 18713284

  • 174.00 € / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRPC5 Monoclonal specifically detects TRPC5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRPC5
Monoclonal
1 mg/mL
Western Blot 1-10 μg/mL, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 0.1-1.0 μg/mL
hTRP5, hTRP-5, short transient receptor potential channel 5, transient receptor potential cation channel, subfamily C, member 5, Transient receptor protein 5, TRP-5, TRP5transient receptor potential channel 5, TrpC5
Mouse
Protein G purified
RUO
Primary
Detects approx 110kDa.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
S67-15
Unconjugated
NP_036603.1
TRPC5
Synthetic peptide amino acids 827-845 of human TrpC5 (also known as short transient receptor potential channel 5, and Htrp5)
0.025 mg
Breast Cancer, Cancer
7224
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only