Learn More
TROY/TNFRSF19 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF1548-SP
Description
TROY/TNFRSF19 Polyclonal specifically detects TROY/TNFRSF19 in Human samples. It is validated for Western Blot.Specifications
| TROY/TNFRSF19 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TAJ, TAJ-alpha, TAJToxicity and JNK inducer, TNFRSF19, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 55504 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| AAF71828 | |
| TNFRSF19 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TROY/TNFRSF19 Glu30-Leu170 Accession # AAF71828 | |
| 25 μg | |
| Primary | |
| Detects human TROY/TNFRSF19 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse (rm) TROY is observed and less than 1% cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, rhNGF R, rhOPG, rhRANK, rhTNF RI, and rhTNF RII is observed. | |
| Human | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.