missing translation for 'onlineSavingsMsg'
Learn More

TROY/TNFRSF19 Antibody (933202), CoraFluor™ 1, Novus Biologicals™

Product Code. 30144650 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30144650 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30144650 Supplier Novus Biologicals Supplier No. FAB15481CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TROY/TNFRSF19 Monoclonal antibody specifically detects TROY/TNFRSF19 in Human samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen TROY/TNFRSF19
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 933202
Conjugate CoraFluor 1
Formulation PBS
Gene Alias TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human TROY/TNFRSF19, Glu30-Leu170, Accession # Q9NS68
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 55504
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.