Learn More
Tropine, TRC
Product Code. 30134809
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134809 50 g 50g
30134832 100 g 100g
17951185 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134809 Supplier Toronto Research Chemicals Supplier No. TRCT89266550G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 120-29-6
Molecular Formula C8 H15 N O
Molecular Weight (g/mol) 141.21
Synonym (1R,3r,5S)-8-Methyl-8-azabicyclo[3.2.1]oct-3-ol, 8-Azabicyclo[3.2.1]octan-3-ol, 8-methyl-, (3-endo)-, 1αH,5αH-Tropan-3α-ol (8CI), 8-Azabicyclo[3.2.1]octan-3-ol, 8-methyl-, endo-, Tropanol (6CI,7CI), (3-endo)-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol, 2,3-Dihydro-3α-hydroxy-8-methylnortropidine, 2,3-Dihydro-3α-hydroxytropidine, 3α-Tropanol, NSC 43870, Tropine, α-Tropine
IUPAC Name (1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-ol
SMILES CN1[C@@H]2CC[C@H]1C[C@@H](O)C2
Specifications
Quantity 50 g
InChI Formula InChI=1S/C8H15NO/c1-9-6-2-3-7(9)5-8(10)4-6/h6-8,10H,2-5H2,1H3/t6-,7+,8+
Formula Weight 141.1154
Chemical Name or Material Tropine