Learn More
BUCHI Trolley, L-200
Shop All Buchi Products
Click to view available options
For Use With (Equipment):
L-200 Freeze Dryer
Unit Size:
Each
Description
- Stainless steel coated
- For use with L-200 Freeze Dryer
Specifications
Specifications
| Capacity (Metric) | >75 kg |
| No. of Shelves | 0 |
| Temperature Range (Metric) | Ambient to -105°C |
| Material | Stainless Steel |
| For Use With (Equipment) | L-200 Freeze Dryer |
| Length (Metric) | 840 mm |
| Weight (English) | 39.7 lb. |
| Weight (Metric) | 18.1 kg |