missing translation for 'onlineSavingsMsg'
Learn More

Tris(cyclopentadienyl)erbium(III), 99% (99.9%-Er) (REO), Thermo Scientific Chemicals

Product Code. p-7029875
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11367189

Brand: Thermo Scientific Alfa Aesar H27389.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39330-74-0
Molecular Formula C15H18Er+3
Molecular Weight (g/mol) 365.568
MDL Number MFCD00145475
InChI Key WEZXZKINGCWZRM-UHFFFAOYSA-N
Synonym erbium 3+ ion tris cyclopenta-1,3-dien-1-ide
PubChem CID 44153880
IUPAC Name cyclopenta-1,3-diene;erbium(3+)
SMILES C1C=CC=C1.C1C=CC=C1.C1C=CC=C1.[Er+3]

Specifications

Melting Point 287°C
Quantity 1 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Formula Weight 362.54
Percent Purity 99%
Assay (99.9%-Er)
Chemical Name or Material Tris(cyclopentadienyl)erbium(III), (REO)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.