missing translation for 'onlineSavingsMsg'
Learn More

Tripelennamine hydrochloride, MedChemExpress

Product Code. 16431679
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
200mg
500mg
5g
This item is not returnable. View return policy

Product Code. 16431679

Brand: Medchem Express HY17428

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tripelennamine hydrochloride, an ethylenediamine derivative, is a potent histamine H1-receptor antagonist. Tripelennamine hydrochloride lessens the allergic response of the organism caused by histamine. Tripelennamine hydrochloride can be used for the research of rhinitis, conjunctivitis, and allergic and anaphylactic reactions.

Tripelennamine hydrochloride, an ethylenediamine derivative, is a potent histamine H1-receptor antagonist. Tripelennamine hydrochloride lessens the allergic response of the organism caused by histamine. Tripelennamine hydrochloride can be used for the research of rhinitis, conjunctivitis, and allergic and anaphylactic reactions.

Specifications

Chemical Name or Material Tripelennamine hydrochloride
CAS 154-69-8
Color White
Purity Grade Notes Research
Molecular Formula C16H22ClN3
Quantity 1 g
Solubility Information H2O : 100 mg/mL (342.68 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (85.67 mM; Need ultrasonic)
SMILES CN(C)CCN(CC1=CC=CC=C1)C2=NC=CC=C2.[H]Cl
Molecular Weight (g/mol) 291.82
Formula Weight 291.82
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.