Learn More
Motic™ Trinocular Head, BA310 MET
Product Code. 15485252
Change view
Click to view available options
For Use With (Equipment):
BA310 MET Microscopes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15485252 BA310 MET Microscopes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15485252 Supplier Motic™ Supplier No. 1101001903241
  • ONLINE EXCLUSIVE PRICE
    537.00 €
    474.96 € / Pack of 1
    Save 62.04 €
    11% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Superior quality Motic™ replacement trinocular, Siedentopf-type microscope head. Motic™ Trinocular Head, BA310 MET is suitable for BA310 MET Microscopes.
  • Siedentopf type.
  • Designed for rBA310 MET Microscopes.
  • Inclined 30°.
Specifications
Type Siedentopf, 30° Incline Trinocular Head
Description Light Split: 100:0/50:50
For Use With (Equipment) BA310 MET Microscopes