Learn More
Trimethoxy Dobutamine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (2RS)-N-[2-(3,4-Dimethoxyphenyl)ethyl]-4-(4-methoxyphenyl)butan-2-amine Hydrochloride |
| CAS | 51062-14-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C21 H29 N O3 . Cl H |
| Quantity | 250 mg |
| Synonym | N-[2-(3,4-Dimethoxyphenyl)ethyl]-4-methoxy-α-methylbenzenepropanamine Hydrochloride (1:1), (2RS)-N-[2-(3,4-Dimethoxyphenyl)-ethyl]-4-(4-methoxyphenyl)-butan-2-amine Hydrochloride, Benzenepropanamine','N-[2-(3,4-dimethoxyphenyl)ethyl]-4-methoxy-α-methyl-','hydrochloride (9CI), Trimethoxy Dobutamine Hydrochloride |
| InChI Formula | InChI=1S/C21H29NO3.ClH/c1-16(5-6-17-7-10-19(23-2)11-8-17)22-14-13-18-9-12-20(24-3)21(15-18)25-4;/h7-12,15-16,22H,5-6,13-14H2,1-4H3;1H |
| SMILES | Cl.COc1ccc(CCC(C)NCCc2ccc(OC)c(OC)c2)cc1 |
| IUPAC Name | N-[2-(3,4-dimethoxyphenyl)ethyl]-4-(4-methoxyphenyl)butan-2-amine;hydrochloride |
| Molecular Weight (g/mol) | 379.9208 |
| Show More |