missing translation for 'onlineSavingsMsg'
Learn More

TRIM38 Antibody, Novus Biologicals™

Product Code. 18231288 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18231288 0.1 mL 0.1mL
18484761 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18231288 Supplier Novus Biologicals Supplier No. NBP187126

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRIM38 Polyclonal specifically detects TRIM38 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM38
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias MGC8946, RING finger protein 15RORETRo/SSA ribonucleoprotein homolog, RNF15, tripartite motif containing 38, tripartite motif-containing 38, tripartite motif-containing protein 38, Zinc finger protein RoRet, zinc-finger protein RoRet
Gene Symbols TRIM38
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:LHEEEKSYLWRLEKEEQQTLSRLRDYEAGLGLKSNELKSHILELEEKCQGSAQKLLQNVNDTLSRSWAVKLETSEAVSLELHTMCNVSKLYFDVKKMLRSHQVSVTLDPDTAHHELILSEDRRQVTRGYTQENQDTSSRRFTA
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 10475
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.