Learn More
Triflusal, TRC
Specifications
Specifications
| Chemical Name or Material | Triflusal |
| CAS | 322-79-2 |
| Molecular Formula | C10 H7 F3 O4 |
| Quantity | 100 mg |
| Synonym | Triflusal, 2-(Acetyloxy)-4-(trifluoromethyl)benzoic acid |
| InChI Formula | InChI=1S/C10H7F3O4/c1-5(14)17-8-4-6(10(11,12)13)2-3-7(8)9(15)16/h2-4H,1H3,(H,15,16) |
| SMILES | CC(=O)Oc1cc(ccc1C(=O)O)C(F)(F)F |
| IUPAC Name | 2-acetyloxy-4-(trifluoromethyl)benzoic acid |
| Molecular Weight (g/mol) | 248.16 |
| Formula Weight | 248.0296 |