missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 2-phosphonobutyrate, 97%

Product Code. p-7045073
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317356 5 g 5g
11327356 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317356 Supplier Thermo Scientific Alfa Aesar Supplier No. L09329.06
Add to basket
This item is not returnable. View return policy

CAS: 17145-91-4 | C10H21O5P | 252.247 g/mol

Reactant used for preparation of furospinosulin-1 and analogs as hypoxia-selective antitumor agents, Aminoquinolines as beta-site amyloid precursor protein cleaving enzyme 1 (BACE1) inhibitors and potential anti-Alzheimer?s drugs, phenylpropanoic acid peroxisome proliferator-activated receptor (PPAR) α-selective agonists as nonalcoholic steatohepatitis (NASH)-preventive agents, PPAR agonists with phenethylphenylphthalimide skeleton derived from thalidomide-related LXR antagonists, notch-sparing γ-secretase inhibitors derived from PPAR agonist library and plakotenin via Diels-Alder reaction, as a potential anticancer agent, naturally found in Okinawan sponge of the genus Plakortis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for preparation of furospinosulin-1 and analogs as hypoxia-selective antitumor agents, Aminoquinolines as beta-site amyloid precursor protein cleaving enzyme 1 (BACE1) inhibitors and potential anti-Alzheimer′s drugs, phenylpropanoic acid peroxisome proliferator-activated receptor (PPAR) α-selective agonists as nonalcoholic steatohepatitis (NASH)-preventive agents, PPAR agonists with phenethylphenylphthalimide skeleton derived from thalidomide-related LXR antagonists, notch-sparing γ-secretase inhibitors derived from PPAR agonist library and plakotenin via Diels-Alder reaction, as a potential anticancer agent, naturally found in Okinawan sponge of the genus Plakortis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17145-91-4
Molecular Formula C10H21O5P
Molecular Weight (g/mol) 252.247
MDL Number MFCD00041347
InChI Key GYUCVQSNZFRDRL-UHFFFAOYSA-N
Synonym triethyl 2-phosphonobutyrate, ethyl 2-diethoxyphosphoryl butanoate, ethyl 2-diethylphosphono butyrate, butanoic acid, 2-diethoxyphosphinyl-, ethyl ester, triethyl 2-phosphonobutanoate, .alpha.-diethylphosphono butanoic acid, ethyl ester, butyric acid, 2-phosphono-, triethyl ester, butyric acid, triethyl ester, triethyl 2-phosphonobu-tyrate, triethyl-2-phosphono butyrate
PubChem CID 229053
IUPAC Name ethyl 2-diethoxyphosphorylbutanoate
SMILES CCC(C(=O)OCC)P(=O)(OCC)OCC

Specifications

Density 1.064
Boiling Point 105°C to 106°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.433
Quantity 5 g
Beilstein 1789280
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 252.25
Percent Purity 97%
Chemical Name or Material Triethyl 2-phosphonobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.