Learn More
Triethyl 1,3,5-benzenetricarboxylate, 97%
Product Code. 11450997
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450997 5 g 5g
11460997 25 g 25g
11470997 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11450997 Supplier Thermo Scientific Alfa Aesar Supplier No. B23392.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 4105-92-4 | C15H18O6 | 294.303 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4105-92-4
Molecular Formula C15H18O6
Molecular Weight (g/mol) 294.303
MDL Number MFCD00236389
InChI Key KXGOWZRHSOJOLF-UHFFFAOYSA-N
Synonym triethyl trimesate, 1,3,5-benzenetricarboxylic acid, triethyl ester, trimesic acid triethyl ester, triethyl 1,3,5-benzenetricarboxylate, ethyl 3,5-bis ethoxycarbonyl benzoate, 1,3,5-triethyl benzene-1,3,5-tricarboxylate, acmc-20a6fr, bzta02, 1,3,5-tri ethoxycarbonyl benzene, triethyl 1,5-benzenetricarboxylate
PubChem CID 226764
IUPAC Name triethyl benzene-1,3,5-tricarboxylate
SMILES CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC)C(=O)OCC
Specifications
Melting Point 132°C to 135°C
Quantity 5 g
Formula Weight 294.31
Percent Purity 97%
Chemical Name or Material Triethyl 1,3,5-benzenetricarboxylate