missing translation for 'onlineSavingsMsg'
Learn More

Triclabendazole, MedChemExpress

Product Code. p-200096376
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16452186

Brand: Medchem Express HYB0621

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Triclabendazole(CGA89317) is a benzimidazole, it binds to tubulin impairing intracellular transport mechanisms and interferes with protein synthesis.

Triclabendazole(CGA89317) is a benzimidazole, it binds to tubulin impairing intracellular transport mechanisms and interferes with protein synthesis.

Specifications

Chemical Name or Material Triclabendazole
CAS 68786-66-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H9Cl3N2OS
Quantity 500 mg
Synonym CGA89317
Solubility Information DMSO : 100 mg/mL (278.04 mM; Need ultrasonic)
SMILES CSC1=NC2=CC(OC3=CC=CC(Cl)=C3Cl)=C(Cl)C=C2N1
Molecular Weight (g/mol) 359.66
Formula Weight 359.66
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.