Learn More
TRIB1 Antibody [PerCP], Novus Biologicals Biologicals™
Product Code. 30500435 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30500435 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30500435 Supplier Novus Biologicals Supplier No. NBP335100PCP
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
TRIB1 Polyclonal antibody specifically detects TRIB1 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen TRIB1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias C8FWG-protein-coupled receptor induced protein, GIG-2, GIG2G-protein-coupled receptor-induced protein 2, G-protein-coupled receptor-induced gene 2 protein, SKIP1, TRB-1, TRB1phosphoprotein regulated by mitogenic pathways, tribbles homolog 1, tribbles homolog 1 (Drosophila), tribbles-like protein 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 203-372 of human TRIB1 (NP_079471.1).,, Sequence:, LGDLKLRKFVFSTEERTQLRLESLEDTHIMKGEDDALSDKHGCPAYVSPEILNTTGTYSGKAADVWSLGVMLYTLLVGRYPFHDSDPSALFSKIRRGQFCIPEHISPKARCLIRSLLRREPSERLTAPEILLHPWFESVLEPGYIDSEIGTSDQIVPEYQEDSDISSFFC
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 10221
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG