Learn More
Tri-tert-butylphosphine, 95%, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 30.0°C |
| Density | 0.8200 g/mL |
| Boiling Point | 102.0°C to 103.0°C (13.0 mmHg) |
| Flash Point | -17°C |
| Assay Percent Range | 95% |
| Packaging | Glass bottle |
| Linear Formula | [(CH3)3C]3P |
| Quantity | 5 g |
| Specific Gravity | 0.82 |
| Solubility Information | Solubility aq. soln.: insoluble aq. soln.. |
| Show More |
Safety and Handling
- Tri-tert-butylphosphine
- Danger
- Flammable solid Category 2
- Pyrophoric solid Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H250-Catches fire spontaneously if exposed to air.
- H228-Flammable solid.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.