missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRF1 Monoclonal Antibody (57-6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA146375
This item is not returnable.
View return policy
Description
This antibody will not work in ICC/IF applications. Suggested positive control: HeLa nuclear extracts, antigen standard for TERF1 (transient overexpression lysate), Hela whole cell extract.
This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products.
Specifications
| TRF1 | |
| Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% sodium azide | |
| P54274, P70371 | |
| TERF1 | |
| Recombinant human TRF1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG1 κ |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| 57-6 | |
| Unconjugated | |
| TERF1 | |
| FLJ41416; hTRF1-AS; NIMA-interacting protein 2; PIN2; Telomeric protein Pin2/TRF1; telomeric repeat binding factor (NIMA-interacting) 1; telomeric repeat binding factor 1; telomeric repeat-binding factor 1; TERF1; TRBF1; TRF; TRF1; TTAGGG repeat-binding factor 1; t-TRF1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 21749, 297758, 7013 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction