Learn More
Masterflex™ Transparent Tygon E-3603 Non-DEHP Tubing
Product Code. 15135403
Change view
Click to view available options
Diameter (Metric) Inner:
1.6 mm
Unit Size:
15m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
15135403 1.6 mm 15m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15135403 Supplier Masterflex™ Supplier No. 0640737
Add to basket
Edge
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Specifications
Certifications/Compliance FDA-compliant (21 CFR 175.300), manufactured according to GMP Compliance
Autoclavable Autoclavable
Diameter (Metric) Inner 1.6 mm
Material Tygon E-3603
Diameter (Metric) Outer 3.2 mm
Overall Length (Metric) 15.24 m
For Use With (Application) General laboratory use
Min. Operating Temperature (Metric) -50°C
Packaging Quantity 1 Pack
Color Transparent
Flexibility Flexible
Max. Operating Temperature (Metric) 74°C