Learn More
Kartell™ Transparent Polypropylene Powder Funnels
Manufactured with polypropylene for strength and durability. Kartell™ Transparent Polypropylene Powder Funnels are designed to funnel large quantities of powders or other solid or liquid materials without flow rate restrictions, powder segregation or bridging.
Brand: Kartell™ 0016900
31.48 EUR valid until 2026-03-31
BEST PRICE on Promo! Use promo code "27994" to get your promotional price.
Description
- 60° angled sides
- External ribbing avoids pressure lock
- Can be used with liquids
- Autoclavable