missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl, 97%

Product Code. p-7036898
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15422066 1 g 1g
15432066 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422066 Supplier Thermo Scientific Alfa Aesar Supplier No. H55356.03
Add to basket
This item is not returnable. View return policy

CAS: 82832-57-3 | C21H30F2 | 320.468 g/mol

It is  used in liquid crystal monomer, pharmaceutical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is  used in liquid crystal monomer, pharmaceutical synthesis.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82832-57-3
Molecular Formula C21H30F2
Molecular Weight (g/mol) 320.468
MDL Number MFCD09838997
InChI Key FSWZOZXLWVWJAH-UHFFFAOYSA-N
Synonym trans,trans-4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 1,2-difluoro-4-4-4-propylcyclohexyl cyclohexyl benzene, trans,trans-4-3,4-difluorophenyl-4-propylbicyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-propyl-bicyclohexyl, 4-3,4-difluoro-phenyl-4'-propyl-bicyclohexyl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexyl, trans,trans-4-3,4-difluorophenyl-4'-n-propylbicyclohexyl, benzene, 1,2-difluoro-4-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4-difluorophenyl-4'-propyl-1,1'-bi cyclohexane, 4-3,4-difluorophenyl-4'-propyl-bicyclohexyl
PubChem CID 581325
IUPAC Name 1,2-difluoro-4-[4-(4-propylcyclohexyl)cyclohexyl]benzene
SMILES CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F

Specifications

Melting Point 43°C to 46°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 320.47
Percent Purity 97%
Chemical Name or Material trans,trans-4-(3,4-Difluorophenyl)-4'-n-propylbicyclohexyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.