Learn More
trans-cis-10,11-Epoxy Farnesenic Acid Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | trans-cis-10,11-Epoxy Farnesenic Acid Methyl Ester |
| CAS | 32466-63-0 |
| Molecular Formula | C16H26O3 |
| Quantity | 2.5 mg |
| Synonym | (Z,E)-9-(3,3-Dimethyloxiranyl)-3,7-dimethyl-2,6-nonadienoic Acid Methyl Ester, Methyl 10,11-Epoxy-(2Z,6E)-farnesate, (Z,E)-10,11-Epoxy-3,7,11-trimethyl-2,6-dodecadienoic Acid, 2,6-Dodecadienoic acid, 10,11-epoxy-3,7,11-trimethyl-, methyl ester, (Z,E)- (8CI), 2,6-Nonadienoic acid, 9-(3,3-dimethyloxiranyl)-3,7-dimethyl-, methyl ester, (Z,E)- (9CI), Methyl (2Z,6E)-9-(3,3-dimethyl-2-oxiranyl)-3,7-dimethyl-2,6-nonadienoate (ACI) |
| InChI Formula | InChI=1S/C16H26O3/c1-12(9-10-14-16(3,4)19-14)7-6-8-13(2)11-15(17)18-5/h7,11,14H,6,8-10H2,1-5H3/b12-7-,13-11+ |
| SMILES | C(C/C(=C/CC/C(=C\C(OC)=O)/C)/C)C1C(C)(C)O1 |
| IUPAC Name | methyl (2Z,6E)-9-(3,3-dimethyloxiran-2-yl)-3,7-dimethylnona-2,6-dienoate |
| Molecular Weight (g/mol) | 266.38 |
| Formula Weight | 266.19 |