Learn More
trans-Cinnamic-alpha,2,3,4,5,6-d6, CDN
Specifications
Specifications
| Chemical Name or Material | trans-Cinnamic Acid D6 (phenyl-D5,alpha-D) |
| CAS | 1174218-97-3 |
| Molecular Formula | C9 D6 H2 O2 |
| Quantity | 0.1 g |
| Synonym | 2-Propenoic-3-d acid, 3-(phenyl-2,3,4,5,6-d5)-, (2 E)- (ACI), (2 E)-3-(Phenyl-2,3,4,5,6-d5)-2-propenoic-3-d acid (ACI), trans-Cinnamic-d6 acid, (E)-3-(Phenyl-d5)-2-propenoic-3-d acid, trans-Cinnamic acid D6 (phenyl-D5,α-D), trans-Cinnamic-α,2,3,4,5,6-d6 acid |
| InChI Formula | InChI=1 S/C9H8O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7 H,(H,10,11)/b7-6+/i1D,2 D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]\C(=C/C(=O)O)\c1c([2 H])c([2 H])c([2 H])c([2 H])c1[2 H] |
| IUPAC Name | (E)-3-deuterio-3-(2,3,4,5,6-pentadeuteriophenyl)prop-2-enoic acid |
| Molecular Weight (g/mol) | 154.196 |
| Formula Weight | 154.09 g/mol |
| Show More |