missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Trifluoromethyl)cinnamic acid, 98%

Product Code. p-7043797
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377835 1 g 1g
11387835 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377835 Supplier Thermo Scientific Alfa Aesar Supplier No. L02771.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16642-92-5 | C10H6F3O2 | 215.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16642-92-5
Molecular Formula C10H6F3O2
Molecular Weight (g/mol) 215.15
MDL Number MFCD00002696
InChI Key ANRMAUMHJREENI-ZZXKWVIFSA-M
Synonym 4-trifluoromethyl cinnamic acid, trans-4-trifluoromethyl cinnamic acid, p-trifluoromethyl cinnamic acid, e-3-4-trifluoromethyl phenyl acrylic acid, 2e-3-4-trifluoromethyl phenyl prop-2-enoic acid, 4-trifluoromethylcinnamic acid, 2-propenoic acid, 3-4-trifluoromethyl phenyl, 3-4-trifluoromethyl-phenyl-acrylic acid, ptf-cna, trans-4-trifluoromethylcinnamic acid
PubChem CID 688070
ChEBI CHEBI:60705
IUPAC Name (E)-3-[4-(trifluoromethyl)phenyl]prop-2-enoic acid
SMILES [O-]C(=O)\C=C\C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 230°C to 234°C
Odor Odorless
Quantity 1 g
Beilstein 2452800
Formula Weight 216.16
Percent Purity 98%
Chemical Name or Material trans-4-(Trifluoromethyl)cinnamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.