Learn More
trans-4-Methycyclohexyl Isocyanate, TRC
Specifications
Specifications
| Chemical Name or Material | trans-4-Methylcyclohexylisocyanate |
| CAS | 32175-00-1 |
| Molecular Formula | C8 H13 N O |
| Quantity | 250 mg |
| Synonym | Cyclohexane','1-isocyanato-4-methyl-','trans-, Isocyanic acid','4-methylcyclohexyl ester','trans- (8CI), trans-1-Isocyanato-4-methylcyclohexane, trans-4-Methylcyclohexyl isocyanate, trans-4-Methylcyclohexylisocyanate |
| InChI Formula | InChI=1S/C8H13NO/c1-7-2-4-8(5-3-7)9-6-10/h7-8H,2-5H2,1H3/t7-,8- |
| SMILES | C[C@@H]1CC[C@H](CC1)N=C=O |
| IUPAC Name | 1-isocyanato-4-methylcyclohexane |
| Molecular Weight (g/mol) | 139.195 |
| Formula Weight | 139.1 |