Learn More
trans-4-Hydroxy-L-proline-d4 (Major), TRC
Specifications
Specifications
| Chemical Name or Material | trans-4-Hydroxy-L-proline-d4 |
| CAS | 2734920-51-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C5H5D4NO3 |
| Quantity | 1 mg |
| Synonym | trans-4-Hydroxy-L-proline-2,5,5-d4, (-)-4-Hydroxy-2-pyrrolidinecarboxylic Acid-d4, (2S,4R)-(-)-4-Hydroxyproline-d4, (R)-4-Hydroxy-(S)-proline-d4 |
| InChI Formula | InChI=1S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7H,1-2H2,(H,8,9)/t3-,4+/m1/s1/i2D2,3D,4D |
| SMILES | O[C@@]1([2H])C([2H])([2H])N[C@]([2H])(C(O)=O)C1 |
| IUPAC Name | (2S,4R)-4-hydroxypyrrolidine-2-carboxylic-2,4,5,5-d4 acid |
| Molecular Weight (g/mol) | 135.15 |
| Show More |