Learn More
trans-4-Hydroxy-3-methoxy-d3-cinnamic Acid, CDN
Product Code. 30319534
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319534 0.005 g 0.01g
30309544 0.01 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319534 Supplier CDN Supplier No. CDND66690.005G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material trans-Ferulic Acid-d3
CAS 860605-59-0
Molecular Formula C10 D3 H7 O4
Quantity 0.005 g
Synonym 2-Propenoic acid, 3-[4-hydroxy-3-(methoxy-d3)phenyl]-, (2 E)- (9 CI), (2 E)-3-(4-Hydroxy-3-[methoxy-d3)phenyl]-2-propenoic acid, (2 E)-3-(4-Hydroxy-3-methoxyphenyl)-2-propenoic acid-d3, (E)-Ferulic-D3 acid (methyl-D3), trans-4-Hydroxy-3-methoxycinnamic acid D3, trans-Ferulic-d3 acid
InChI Formula InChI=1 S/C10H10O4/c1-14-9-6-7(2-4-8(9)11)3-5-10(12)13/h2-6,11 H,1H3,(H,12,13)/b5-3+/i1D3
SMILES [2 H]C([2 H])([2 H])Oc1cc(\C=C\C(=O)O)ccc1O
IUPAC Name (E)-3-[4-hydroxy-3-(trideuteriomethoxy)phenyl]prop-2-enoic acid
Molecular Weight (g/mol) 197.202
Formula Weight 197.077 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity