Learn More
trans-4-Acetamidocyclohexanol, TRC
Specifications
Specifications
| Chemical Name or Material | trans-4-Acetamidocyclohexanol |
| CAS | 27489-60-7 |
| Molecular Formula | C8H15NO2 |
| Quantity | 1 g |
| Synonym | trans-N-(4-Hydroxycyclohexyl)-acetamide, trans-N-Acetyl-4-hydroxycyclohexylamine |
| InChI Formula | InChI=1S/C8H15NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h7-8,11H,2-5H2,1H3,(H,9,10) |
| SMILES | CC(=O)N[C@@H]1CC[C@@H](O)CC1 |
| IUPAC Name | N-(4-hydroxycyclohexyl)acetamide |
| Molecular Weight (g/mol) | 157.21 |
| Formula Weight | 157.1103 |