missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid, 98%

Product Code. p-7032775
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418915 5 g 5g
15428915 25 g 25g
15438915 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15418915 Supplier Thermo Scientific Alfa Aesar Supplier No. H34384.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 49708-81-8 | C13H15ClO2 | 238.711 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49708-81-8
Molecular Formula C13H15ClO2
Molecular Weight (g/mol) 238.711
MDL Number MFCD06797626
InChI Key NXXDIEYTMQYWJU-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl cyclohexanecarboxylic acid, trans-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4-chlorophenyl cyclohexane-1-carboxylic acid, e-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4'-chlorophenyl cyclohexancarboxylic acid, cis-4-4-chlorophenyl cyclohexanecarboxylic acid, atovaquone related compound 1, cyclohexanecarboxylic acid, 4-4-chlorophenyl-, trans, 1r,4r-4-4-chlorophenyl cyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 4-4-chlorophenyl
PubChem CID 2735792
IUPAC Name 4-(4-chlorophenyl)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C2=CC=C(C=C2)Cl)C(=O)O

Specifications

Melting Point 249°C to 254°C
Quantity 5 g
Formula Weight 238.71
Percent Purity 98%
Chemical Name or Material trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.