missing translation for 'onlineSavingsMsg'
Learn More

trans-1,4-Cyclohexanedicarboxylic acid, 95%

Product Code. 15486558
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15486558 5 g 5g
15476558 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15486558 Supplier Thermo Scientific Alfa Aesar Supplier No. H60089.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 619-82-9 | C8H12O4 | 172.18 g/mol

It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate.

Solubility
Soluble in hot methanol (almost transparency), ethanol, acetone. Slightly soluble in ether and water

Notes
Store in cool dry conditions. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-82-9
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00066203
InChI Key PXGZQGDTEZPERC-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedicarboxylic acid, trans-1,4-cyclohexanedicarboxylic acid, cis-1,4-cyclohexanedicarboxylic acid, cis-cyclohexane-1,4-dicarboxylic acid, trans-cyclohexane-1,4-dicarboxylic acid, hexahydroterephthalic acid, trans-hexahydroterephthalic acid, trans-1,4-cyclohexanedicarboxybic acid, cis-hexahydroterephthalic acid, 1,4-dicarboxycyclohexane
PubChem CID 14106
IUPAC Name cyclohexane-1,4-dicarboxylic acid
SMILES C1CC(CCC1C(=O)O)C(=O)O

Specifications

Melting Point >300°C
Quantity 5 g
Solubility Information Soluble in hot methanol (almost transparency),ethanol,acetone. Slightly soluble in ether and water
Formula Weight 172.18
Percent Purity 95%
Chemical Name or Material trans-1,4-Cyclohexanedicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.