Learn More
trans-1,2,3,4,4a,10,11,11a-Octahydro-N,N-dimethyl-5H-Dibenzo[a,d]cycloheptene-Delta5,Gamma-propylamine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | trans-1,2,3,4,4a,10,11,11a-Octahydro-N,N-dimethyl-5H-Dibenzo[a,d]cycloheptene-delta5,gamma-propylamine Hydrochloride |
| CAS | 27867-02-3 |
| Molecular Formula | C20 H29 N . H Cl |
| Quantity | 10 mg |
| Synonym | Amitriptyline Impurity E, Amitriptyline EP Impurity E |
| InChI Formula | InChI=1S/C20H29N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3,5,8,10,12,17,19H,4,6-7,9,11,13-15H2,1-2H3;1H/t17-,19+;/m1./s1 |
| SMILES | [H][C@]1(CCC2=C/3C=CC=C2)[C@](CCCC1)([H])C3=C/CCN(C)C.Cl |
| IUPAC Name | 3-[(4aR,11aS)-1,2,3,4,4a,5,6,11a-octahydrodibenzo[2,1-a:1',2'-d][7]annulen-11-ylidene]-N,N-dimethylpropan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 319.91 |
| Formula Weight | 319.2067 |