Learn More
Tranexamic Acid, TRC
Product Code. 17942985
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942985 1 g 1g
17961575 10 g 10g
17921685 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17942985 Supplier Toronto Research Chemicals Supplier No. TRCT7145051G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1197-18-8
Molecular Formula C8 H15 N O2
Molecular Weight (g/mol) 157.21
Synonym Cyclohexanecarboxylic acid, 4-(aminomethyl)-, trans-, trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 4-trans-Aminomethylcyclohexanecarboxylic acid, AMCA, AMCHA, Amchafibrin, Amikapron, Anvitoff, Bay 3517, CL 65336, Cyclocapron, Cyclokapron, Cyklokapron, DV 79, Exacyl, HAKU, Haematrix, Lysteda, TAMCHA, Tranexamic acid, Tranol, Transamin, Transamin G, Transamin S, Transcam, Ugurol, Xamig, Xed, Zataranax, t-AMCHA, trans-4-(Aminomethyl)-1-cyclohexanecarboxylic acid, trans-Amcha, trans-Tranexamic acid, trans-p-(Aminomethyl)cyclohexanecarboxylic acid
IUPAC Name 4-(aminomethyl)cyclohexane-1-carboxylic acid
SMILES NC[C@@H]1CC[C@H](CC1)C(=O)O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C8H15NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h6-7H,1-5,9H2,(H,10,11)/t6-,7-
Formula Weight 157.1103
Percent Purity >95%
Chemical Name or Material Tranexamic acid
Grade HPLC