Learn More
TRAF4 Mouse anti-Human, CoraLite 488, Clone: 2C9A11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66755-100UL
Description
The TRAF4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRAF4. This antibody recognizes human antigen. The TRAF4 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| TRAF4 | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 9618 | |
| IgG1 |
| Immunofluorescence | |
| 2C9A11 | |
| Mouse | |
| Primary | |
| Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.