missing translation for 'onlineSavingsMsg'
Learn More

TRAF3IP2 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18728211 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18728211 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18728211 Supplier Novus Biologicals Supplier No. NB10093556AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRAF3IP2 Polyclonal antibody specifically detects TRAF3IP2 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAF3IP2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias ACT1DKFZP586G0522, adapter protein CIKS, C6orf2, C6orf4chromosome 6 open reading frame 5, C6orf5DKFZp586G0522, C6orf6MGC3581, CIKSchromosome 6 open reading frame 2, Connection to IKK and SAPK/JNK, NFkB-activating protein ACT1, Nuclear factor NF-kappa-B activator 1, TRAF3 interacting protein 2, TRAF3-interacting protein 2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminus of human ACT.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10758
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.