missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRA-1-60 Monoclonal Antibody (TRA-1-60), PE

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA1023PE
This item is not returnable.
View return policy
Description
MA1-023-PE has been successfully used in ICC/IF and flow cytometry applications on human samples.
TRA-1-60 is a cell surface antigen, expressed along with SSEA-3, SSEA-4 and TRA-1-81 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are lost during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
Specifications
| TRA-1-60 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| Mouse | |
| Affinity chromatography - MBP | |
| RUO | |
| Human | |
| Antibody | |
| IgM |
| Flow Cytometry | |
| TRA-1-60 | |
| PE | |
| TRA160; tra60 | |
| Human embryonal carcinoma cell line 2102Ep. | |
| 100 μL | |
| Primary | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction