missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TR beta 1/NR1A2/Thyroid Hormone Receptor beta Antibody (2386), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-61088
This item is not returnable.
View return policy
Description
TR beta 1/NR1A2/Thyroid Hormone Receptor beta Monoclonal specifically detects TR beta 1/NR1A2/Thyroid Hormone Receptor beta in Human samples. It is validated for Western Blot.
Specifications
| TR beta 1/NR1A2/Thyroid Hormone Receptor beta | |
| Monoclonal | |
| Unconjugated | |
| P10828 | |
| THRB | |
| Synthetic peptide corresponding to amino acid residues from the N-terminal region conjugated to KLH. Accession # P10828 | |
| Protein G purified | |
| RUO | |
| Primary | |
| Specific for the ∼55k TR-beta protein. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| 2386 | |
| Western Blot 1:1000 | |
| avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, c-erbA-2, c-erbA-beta, ERBA2MGC126109, ERBA-BETA, GRTH, MGC126110, NR1A2thyroid hormone receptor, beta (avian erythroblastic leukemia viral (v-erb-a)oncogene homolog 2), Nuclear receptor subfamily 1 group A member 2, oncogene ERBA2, PRTH, THR1pituitary resistance to thyroid hormone, THRB1, THRB2, thyroid hormone receptor beta, thyroid hormone receptor beta 1, thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a)oncogene homolog 2, avian) | |
| Mouse | |
| 55 kDa | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 7068 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction