missing translation for 'onlineSavingsMsg'
Learn More

TR-14035, MedChemExpress

Product Code. 16426818
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16426818

Brand: Medchem Express HY15770

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TR-14035 is a orally active dual α4β7/α4β1 integrin antagonist, with IC50 s of 7 nM and 87 nM for α4β7 and α4β1, respectively. TR-14035 can be used for the research of inflammation and autoimmune diseases.

TR-14035 is a orally active dual α4β7/α4β1 integrin antagonist, with IC50 s of 7 nM and 87 nM for α4β7 and α4β1, respectively. TR-14035 can be used for the research of inflammation and autoimmune diseases.

Specifications

Chemical Name or Material TR-14035
CAS 232271-19-1
Color White
Purity Grade Notes Research
Molecular Formula C24H21Cl2NO5
Quantity 10 mg
Solubility Information DMSO : ≥ 41 mg/mL (86.44 mM)
SMILES COC1=C(C2=CC=C(C[C@@H](C(O)=O)NC(C3=C(Cl)C=CC=C3Cl)=O)C=C2)C(OC)=CC=C1
Molecular Weight (g/mol) 474.33
Formula Weight 474.33
Percent Purity 95.81%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.