Learn More
Epredia™ TPX Single Sample Chamber, Caps and Filter Cards
For use with disposable white filter cards for sample volumes up to 0.5 mL and for use with disposable brown filter cards for sample volumes of 0.4 mL or less.
Brand: Epredia™ 5991022
Additional Details : Weight : 0.18000kg
Description
TPX Single Sample Chamber
- Reusable
- Autoclavable
- Highly absorbent disposable filter cards for use with TPX chamber
- Not for use with Cytofunnel™ disposable sample chambers
Specifications
White Filter Cards | |
Single-use | |
Sample Chamber, Caps, Filter Card | |
0.5 mL |