Learn More
TPPB, MedChemExpress
Product Code. 16481239
Encompass_Preferred
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 16481239
Brand: Medchem Express HY12359
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress TPPB is a cell-permeable benzolactam-derived protein kinase C (PKC) activator with a Ki of 11.9 nM.
TPPB is a cell-permeable benzolactam-derived protein kinase C (PKC) activator with a Ki of 11.9 nM.
Specifications
Chemical Name or Material TPPB
Physical Form Solid
CAS 497259-23-1
Color Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C27H30F3N3O3
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information DMSO : 125 mg/mL (249.23 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(N(C)[C@@H](C(C)C)C(N[C@H](CO)C2)=O)C2=C1)/C=C/C=C/C3=CC=C(C(F)(F)F)C=C3
Molecular Weight (g/mol) 501.54
Formula Weight 501.54
Percent Purity 99.12%
Grade Research